Learn More
Cole-Parmer™ SH-200 Series Reciprocating Shakers
Product Code. 17701288 Shop All Cole-Parmer Products
Change view
Click to view available options
Speed Range:
25 to 250 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
17701288 25 to 250 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17701288 Supplier Cole-Parmer™ Supplier No. 5190102
  • ONLINE EXCLUSIVE PRICE
    21625.00 NOK
    20490.95 NOK / Pack of 1
    Save 1134.05 NOK
    5% Off
View all promotions
Estimated Shipment: 03-12-2026
Powerful yet quiet, perfect for continuous use applications.
  • Digital speed selection provides consistent mixing
  • Soft-start motor prevents spills and splashes
  • Fully adjustable cradle accommodates most flasks, bottles, and beakers
  • Can be used in incubators system
These linear reciprocating shakers feature digital speed selection from 25 to 250 rpm with a soft start. Four adjustable rubber cushioned horizontal securing bars with quick release handles on the cradle-type platform allow you to fit nearly any size flask, bottle, or beaker. Secure up to twelve 250 mL, nine 500 mL, four 1 L, or two 2 L vessels. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Shaker is capable of continuous use, and the microprocessor maintains speed accuracy over long runs.
Specifications
Load Bearing Capacity (Metric) 10 kg
Speed Range 25 to 250 rpm
Voltage 230 VAC
Load Bearing Capacity (English) 22 lb.
Frequency 50 Hz
Depth (English) 17.716 in.
Depth (Metric) 45 cm
Dimensions (LxW) Platform 33.5 x 33.5 cm (13 x 13 in.)
Height (English) 10.82 in.
Height (Metric) 27.5 cm
Length (English) Stroke 0.75 in.
Length (Metric) Stroke 20 mm
Width (English) 15.35 in.
Width (Metric) 39 cm
Description Large Reciprocating Shaker
Display LED
Timer Range 1 to 999 rpm