Learn More
SGTB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15895-1-AP-150UL
This item is not returnable.
View return policy
Description
The SGTB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SGTB. This antibody recognizes human, mouse, rat antigen. The SGTB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SGTB | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 54557 | |
| IgG |
| Western Blot | |
| 133 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |