Learn More
SGMS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19050-1-AP-20UL
 View more versions of this product
Product Code. 16818844
  • 1710.33 NOK / 20µL
Description
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
Specifications
Specifications
SGMS1
Polyclonal
Unconjugated
SGMS1
hmob33, MOB, MOB1, Protein Mob, SGMS1, SMS1, sphingomyelin synthase 1, TMEM23, Transmembrane protein 23
Rabbit
Antigen Affinity Chromatography
RUO
208449, 259230, 353229
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TSX5, Q86VZ5, Q8VCQ6
Sgms1
SGMS1 Fusion Protein Ag5246
20 μL
Primary
Mouse, Rat, Human