Learn More
SGLT2/SLC5A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-92384
 View more versions of this product
Product Code. 18776693
  • ONLINE EXCLUSIVE PRICE
    5065.00 NOK
    4791.15 NOK / 0.10mL
    Save 273.85 NOK
    5% Off
Description
SGLT2/SLC5A2 Polyclonal antibody specifically detects SGLT2/SLC5A2 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry, Immunohistochemistry (Paraffin)
This antibody was developed against Recombinant Protein corresponding to amino acids:FHEVGGYSGLFDKYLGAATSLTVSEDPAVGNISSFCYRPRPDSYHLL
RUO
Rabbit
0.1 mL
Certificates
Promotions
Promotions
For Research Use Only