missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11185-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a serine/threonine protein kinase. Although this gene product is similar to serum- and glucocorticoid-induced protein kinase (SGK), this gene is not induced by serum or glucocorticoids. This gene is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Two alternate transcripts encoding two different isoforms have been described.Specifications
| SGK2 | |
| Polyclonal | |
| Unconjugated | |
| SGK2 | |
| H SGK2, SGK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10110, 171497, 27219 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8R4U9, Q9HBY8, Q9QZS5 | |
| Sgk2 | |
| SGK2 Fusion Protein Ag1669 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction