missing translation for 'onlineSavingsMsg'
Learn More

SGK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16848093 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16848093 20 μL 20µL
16838093 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16848093

Brand: Proteintech 111851AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a serine/threonine protein kinase. Although this gene product is similar to serum- and glucocorticoid-induced protein kinase (SGK), this gene is not induced by serum or glucocorticoids. This gene is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Two alternate transcripts encoding two different isoforms have been described.

Specifications

Antigen SGK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SGK2
Gene Accession No. Q8R4U9, Q9HBY8, Q9QZS5
Gene Alias H SGK2, SGK2
Gene Symbols Sgk2
Host Species Rabbit
Immunogen SGK2 Fusion Protein Ag1669
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10110, 171497, 27219
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.