Learn More
Trajan™ Varian/Bruker Replacement Needle
Product Code. 15248593
Click to view available options
Needle Gauge:
23 G
25 G
Diameter (Metric) Outer:
0.5 mm
0.63 mm
Unit Size:
Each
Pack of 2
Product Code. 15248593
Brand: Trajan™ 037779
Autosampler syringe designed for optimal sample introduction. For use with Bruker/Varian GC Systems
  • For use with Bruker/Varian GC Systems with
    • Needle Length: 53mm
    • Tip style: Side-hole
Specifications
Needle Gauge 23 G
Diameter (Metric) Outer 0.63 mm
Product Type Replacement Needle Varian/Bruker
Needle Point Style S-hole
For Use With (Equipment) Autosampler Syringes, 8035, 8100 and 8200