Learn More
SGE™ Tubing Cutter
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- Made of stainless-steel
- O.D.: 1/8 in.
Specifications
Specifications
| Product Type | Tubing Cutter |
| For Use With (Equipment) | GC tubing |
| Material | Stainless Steel |
| For Use With (Application) | Used to get right angle cuts with minimal burring or chipping of the tubing |