Learn More
Trajan™ Thermo Scientific AS3000, AS2000 and AS200/800GC Autosampler Syringe
Fixed needle autosampler syringes. For reduced non specific sample interaction and increased analyte recovery
Brand:  Trajan™ 0029801
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 16682572
  • 695.00 NOK / Each
Estimated Shipment: 27-06-2024
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
26 G
AS3000, AS2000 and AS200/800GC Autosampler Syringe
10 μL
0.47 mm
1/Pk.