Learn More
Trajan™ SuperFlex™ Microsyringes
Description
- ±1% Accuracy and reproducibility
- Ideal for making microvolume injections of 1.5μL to 8μL.
- Each plunger is individually fitted for each microsyringe for perfect sealing
- Plunger protection incorporates a plunger protection guide and plunger button reinforcement to minimize the risk of bent plungers.
- Guided plunger microsyringes have an extended barrel and reinforced plunger that provides additional protection.
Specifications
Specifications
| Material | SuperfleX (Plunger) |
| Needle Gauge | 26 G |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Volume (Metric) | 10 μL |
| Length (Metric) Needle | 50 mm |
| Needle Point Style | Cone |
| Connection Type | Fixed |