Learn More
Trajan™ SI-7 Solids Syringe
Description
- For use with Pyrolysis chromatography applications
- Works in conjunction with Pyrojector II which thermally fragments essentially non-volatile molecules into volatile fragments, making them suitable for chromatographic analysis
Specifications
Specifications
| Material | Stainless Steel |
| Needle Gauge | 25 G |
| For Use With (Application) | For solids injection on SGE pyrojector |
| Length (English) | 2.75 in. |
| Length (English) Needle | 2.75 in. |
| Length (Metric) | 70 mm |
| Product Type | Solids Syringe |