Learn More
- Home
- Products
- Chromatography Syringes
- Chromatography Syringe Accessories
- Trajan™ Replacement Plungers
Trajan™ Replacement Plungers
Product Code. 15298543
Change view
Click to view available options
For Use With (Equipment):
10 μL Gas-tight Autosampler Fixed Needle Syringe
10 μL Removable Guided Plunger Syringe
10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler
100 μL Fixed Needle Gas-tight Syringe for Carlo Erba, CTC, Fisons Autosamplers
Fixed Needle Gas Tight Syringe (10μL)
P250-GT Code Syringe (250μL)
Removable Fitting Gas Tight Syringe (10 mL)
Removable Needle Gas Tight Syringe (10μL)
Rheodyne and Valco Valves
Syringe 009472, 009462
Syringe 100μL
Syringe Models SG-003987, SG-003988, 25 μL
Syringe Models SG-003988, SG-003985, 25 μL
Syringe Needles (10 μL)
Syringe Needles (10 μL), Fixed Needle Gas Tight Syringe
Syringe Needles (100μL), Gas Tight Syringe
Syringe Needles (100μL), Removable Needle Gas Tight Syringe
Syringe Needles (25μL), CTC Gas Tight Syringe
Syringe Needles (25μL), Fixed Needle Gas Tight Syringe
Syringe Needles (25μL), Fixed or Removable Needle Gas Tight Syringe
Syringe Needles (250μL), CTC Gas Tight Syringe
Syringe Needles (250μL), Gas Tight Syringe
Syringe Needles (5 mL), Gas Tight Syringe
Syringe Needles (5μL), Fixed Needle Gas Tight Syringe
Syringe Needles (50μL), Fixed Needle Gas Tight Syringe
Syringe Needles (500μL), CTC Gas Tight Syringe
Syringe Needles (500μL), Fixed or Removable Needle Gas Tight Syringe
Product Type:
50 mL Replacement Plunger only
Plunger
Replacement Plunger (old style), P25F-BT-GT
Replacement Plunger, 10A-VP
Replacement Plunger, 10R-C/T-GT
Replacement Plunger, P10-GP-GT
Replacement Plunger, P100-GT
Replacement Plunger, P100F-C/T-GT
Replacement Plunger, P100R-C/T-GT
Replacement Plunger, P10F-BT-GT
Replacement Plunger, P10F-C/T-GT
Replacement Plunger, P10MD-GT
Replacement Plunger, P10R-GT
Replacement Plunger, P25-GT
Replacement Plunger, P250F-CTC-GT
Replacement Plunger, P250R-C/T-GT
Replacement Plunger, P25F-C/T-GT
Replacement Plunger, P25F-CTC-GT
Replacement Plunger, P25MD-GT
Replacement Plunger, P25R-C/T-GT
Replacement Plunger, P500-GT
Replacement Plunger, P500F-CTC-GT
Replacement Plunger, P50F-C/T-GT
Replacement Plunger, P5F-PE-GT
Replacement Plunger, P5MD-GT
Unit Size:
Each
Pack of 2
| Product Code. | For Use With (Equipment) | Product Type | unitSize |
|---|---|---|---|
| 15298543 | Syringe 100μL | Plunger | Each |
| 15208533 | 10 μL Removable Guided Plunger Syringe | Replacement Plunger, P10-GP-GT | Each |
| 15248573 | Syringe Needles (500μL), CTC Gas Tight Syringe | Replacement Plunger, P500F-CTC-GT | Each |
| 15258533 | 10 μL Gas-tight Autosampler Fixed Needle Syringe | Replacement Plunger, P10F-C/T-GT | Pack of 2 |
| 10274982 | Syringe Needles (10 μL) | Replacement Plunger, 10A-VP | Each |
| 15298523 | Syringe Needles (10 μL), Fixed Needle Gas Tight Syringe | Replacement Plunger, P10F-C/T-GT | Each |
| 12801901 | Syringe Needles (5μL), Fixed Needle Gas Tight Syringe | Replacement Plunger, P5F-PE-GT | Each |
| 15218543 | Syringe Models SG-003987, SG-003988, 25 μL | Replacement Plunger, P25F-C/T-GT | Each |
| 15298533 | Syringe Needles (25μL), Fixed or Removable Needle Gas Tight Syringe | Replacement Plunger, P25-GT | Each |
| 15248533 | Removable Needle Gas Tight Syringe (10μL) | Replacement Plunger, P10R-GT | Pack of 2 |
| 12841901 | Syringe Needles (100μL), Gas Tight Syringe | Replacement Plunger, P100-GT | Each |
| 10388254 | Syringe Needles (100μL), Removable Needle Gas Tight Syringe | Replacement Plunger, P100R-C/T-GT | Each |
| 15288563 | Removable Fitting Gas Tight Syringe (10 mL) | Replacement Plunger, P10MD-GT | Each |
| 15268563 | Syringe Needles (5 mL), Gas Tight Syringe | Replacement Plunger, P5MD-GT | Each |
| 10424540 | Syringe Needles (500μL), Fixed or Removable Needle Gas Tight Syringe | Replacement Plunger, P500-GT | Each |
| 15208573 | Syringe 009472, 009462 | Replacement Plunger, P25MD-GT | Each |
| 15238573 | Syringe Needles (250μL), CTC Gas Tight Syringe | Replacement Plunger, P250F-CTC-GT | Each |
| 15228573 | Syringe Needles (25μL), CTC Gas Tight Syringe | Replacement Plunger, P25F-CTC-GT | Each |
| 15208543 | Syringe Models SG-003988, SG-003985, 25 μL | Replacement Plunger, P25R-C/T-GT | Each |
| 15288543 | 100 μL Fixed Needle Gas-tight Syringe for Carlo Erba, CTC, Fisons Autosamplers | Replacement Plunger, P100F-C/T-GT | Each |
| 15278533 | 10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler | Replacement Plunger, 10R-C/T-GT | Pack of 2 |
| 15268543 | Syringe Needles (50μL), Fixed Needle Gas Tight Syringe | Replacement Plunger, P50F-C/T-GT | Each |
| 15238553 | Syringe Needles (250μL), Gas Tight Syringe | Replacement Plunger, P250R-C/T-GT | Each |
| 15268533 | Fixed Needle Gas Tight Syringe (10μL) | Replacement Plunger, P10F-BT-GT | Each |
| 15228543 | Syringe Needles (25μL), Fixed Needle Gas Tight Syringe | Replacement Plunger (old style), P25F-BT-GT | Each |
| 10525749 | P250-GT Code Syringe (250μL) | Plunger | Each |
| 10897130 | Rheodyne and Valco Valves | 50 mL Replacement Plunger only | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- For use with SGE gas tight syringes.
- Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Specifications
| Product Type | Plunger |
| For Use With (Equipment) | Syringe 100μL |