Learn More
Trajan™ Replacement Plungers
Product Code. 15298523
Change view
Click to view available options
For Use With (Equipment):
10 μL Gas-tight Autosampler Fixed Needle Syringe
10 μL Removable Guided Plunger Syringe
10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler
100 μL Fixed Needle Gas-tight Syringe for Carlo Erba, CTC, Fisons Autosamplers
Fixed Needle Gas Tight Syringe (10μL)
P250-GT Code Syringe (250μL)
Removable Fitting Gas Tight Syringe (10 mL)
Removable Needle Gas Tight Syringe (10μL)
Rheodyne and Valco Valves
Syringe 009472, 009462
Syringe 100μL
Syringe Models SG-003987, SG-003988, 25 μL
Syringe Models SG-003988, SG-003985, 25 μL
Syringe Needles (10 μL)
Syringe Needles (10 μL), Fixed Needle Gas Tight Syringe
Syringe Needles (100μL), Gas Tight Syringe
Syringe Needles (100μL), Removable Needle Gas Tight Syringe
Syringe Needles (25μL), CTC Gas Tight Syringe
Syringe Needles (25μL), Fixed Needle Gas Tight Syringe
Syringe Needles (25μL), Fixed or Removable Needle Gas Tight Syringe
Syringe Needles (250μL), CTC Gas Tight Syringe
Syringe Needles (250μL), Gas Tight Syringe
Syringe Needles (5 mL), Gas Tight Syringe
Syringe Needles (5μL), Fixed Needle Gas Tight Syringe
Syringe Needles (50μL), Fixed Needle Gas Tight Syringe
Syringe Needles (500μL), CTC Gas Tight Syringe
Syringe Needles (500μL), Fixed or Removable Needle Gas Tight Syringe
Product Type:
50 mL Replacement Plunger only
Plunger
Replacement Plunger (old style), P25F-BT-GT
Replacement Plunger, 10A-VP
Replacement Plunger, 10R-C/T-GT
Replacement Plunger, P10-GP-GT
Replacement Plunger, P100-GT
Replacement Plunger, P100F-C/T-GT
Replacement Plunger, P100R-C/T-GT
Replacement Plunger, P10F-BT-GT
Replacement Plunger, P10F-C/T-GT
Replacement Plunger, P10MD-GT
Replacement Plunger, P10R-GT
Replacement Plunger, P25-GT
Replacement Plunger, P250F-CTC-GT
Replacement Plunger, P250R-C/T-GT
Replacement Plunger, P25F-C/T-GT
Replacement Plunger, P25F-CTC-GT
Replacement Plunger, P25MD-GT
Replacement Plunger, P25R-C/T-GT
Replacement Plunger, P500-GT
Replacement Plunger, P500F-CTC-GT
Replacement Plunger, P50F-C/T-GT
Replacement Plunger, P5F-PE-GT
Replacement Plunger, P5MD-GT
Unit Size:
Each
Pack of 2
Product Code. For Use With (Equipment) Product Type unitSize
15298523 Syringe Needles (10 μL), Fixed Needle Gas Tight Syringe Replacement Plunger, P10F-C/T-GT Each
15208533 10 μL Removable Guided Plunger Syringe Replacement Plunger, P10-GP-GT Each
15248573 Syringe Needles (500μL), CTC Gas Tight Syringe Replacement Plunger, P500F-CTC-GT Each
15258533 10 μL Gas-tight Autosampler Fixed Needle Syringe Replacement Plunger, P10F-C/T-GT Pack of 2
10274982 Syringe Needles (10 μL) Replacement Plunger, 10A-VP Each
12801901 Syringe Needles (5μL), Fixed Needle Gas Tight Syringe Replacement Plunger, P5F-PE-GT Each
15218543 Syringe Models SG-003987, SG-003988, 25 μL Replacement Plunger, P25F-C/T-GT Each
15298533 Syringe Needles (25μL), Fixed or Removable Needle Gas Tight Syringe Replacement Plunger, P25-GT Each
15248533 Removable Needle Gas Tight Syringe (10μL) Replacement Plunger, P10R-GT Pack of 2
12841901 Syringe Needles (100μL), Gas Tight Syringe Replacement Plunger, P100-GT Each
10388254 Syringe Needles (100μL), Removable Needle Gas Tight Syringe Replacement Plunger, P100R-C/T-GT Each
15288563 Removable Fitting Gas Tight Syringe (10 mL) Replacement Plunger, P10MD-GT Each
15268563 Syringe Needles (5 mL), Gas Tight Syringe Replacement Plunger, P5MD-GT Each
10424540 Syringe Needles (500μL), Fixed or Removable Needle Gas Tight Syringe Replacement Plunger, P500-GT Each
15208573 Syringe 009472, 009462 Replacement Plunger, P25MD-GT Each
15238573 Syringe Needles (250μL), CTC Gas Tight Syringe Replacement Plunger, P250F-CTC-GT Each
15228573 Syringe Needles (25μL), CTC Gas Tight Syringe Replacement Plunger, P25F-CTC-GT Each
15208543 Syringe Models SG-003988, SG-003985, 25 μL Replacement Plunger, P25R-C/T-GT Each
15288543 100 μL Fixed Needle Gas-tight Syringe for Carlo Erba, CTC, Fisons Autosamplers Replacement Plunger, P100F-C/T-GT Each
15278533 10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler Replacement Plunger, 10R-C/T-GT Pack of 2
15268543 Syringe Needles (50μL), Fixed Needle Gas Tight Syringe Replacement Plunger, P50F-C/T-GT Each
15238553 Syringe Needles (250μL), Gas Tight Syringe Replacement Plunger, P250R-C/T-GT Each
15268533 Fixed Needle Gas Tight Syringe (10μL) Replacement Plunger, P10F-BT-GT Each
15228543 Syringe Needles (25μL), Fixed Needle Gas Tight Syringe Replacement Plunger (old style), P25F-BT-GT Each
10525749 P250-GT Code Syringe (250μL) Plunger Each
15298543 Syringe 100μL Plunger Each
10897130 Rheodyne and Valco Valves 50 mL Replacement Plunger only Each
27 options
Product Code. 15298523
Brand: Trajan™ 031803
Replacement Plungers are available for all SGE gas tight syringes.
  • For use with SGE gas tight syringes.
  • Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Material PTFE (Tip)
For Use With (Application) IN VITRO use only. For lab analyzers only. Not for human use.
Product Type Replacement Plunger, P10F-C/T-GT
For Use With (Equipment) Syringe Needles (10 μL), Fixed Needle Gas Tight Syringe