Learn More
SGE™ Replacement Needles for Removable Needle Syringes
Product Code. 13417248
Change view
Click to view available options
Needle Gauge:
23 G
25 G
Diameter (English) Outer Needle:
0.019 in.
0.024 in.
Unit Size:
Pack of 2
Pack of 5
Product Code. Needle Gauge Diameter (English) Outer Needle unitSize
13417248 23 G 0.024 in. Pack of 5
15238593 25 G 0.019 in. Pack of 2
2 options
Product Code. 13417248
Brand: SGE™ 037111
Replacement needles as supplied with syringe
  • For 10μL syringes
  • 50mm (2 in.) exposed length when fitted to syringe
Specifications
Needle Gauge 23 G
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer 0.63 mm
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Length (English) 2 in.
Length (English) Needle 1.9 in.
Length (Metric) 50 mm
Product Type Replacement Needle
Quantity Ea.
Volume (Metric) 10 μL
Needle Point Style Bevel
For Use With (Equipment) Removable Needle Syringe (10μL)