Learn More
Trajan™ Replacement Needles for 25 to 500μL Removable Needle Syringes
Description
Replacement Needles for 25 to 500uL Syringes for use in Chromatography
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
| Material | Stainless Steel |
| Needle Gauge | 23 G |
| Diameter (English) Inner | 0.009 in. |
| Diameter (English) Outer | 0.024 in. |
| Diameter (Metric) Inner | 0.24 mm |
| Diameter (Metric) Outer | 0.63 mm |
| For Use With (Application) | Chromatography |
| Length (English) Needle | 4.52 in. |
| Product Type | Replacement Needle |
| Quantity | 5/Pk. |
| Show More |