Learn More
Trajan™ Replacement Kit for Plunger-in-Needle Syringes
Click to view available options
Diameter (Metric) Outer Needle:
0.47 mm
0.63 mm
For Use With (Equipment):
6.5 mm OD Syringe (0.5μL)
8 mm OD Syringe (1μL)
Syringes (1μL)
Unit Size:
Each
Description
- High precision and accuracy for small volume injections
- Replacement plunger/needle kits
- Kits are matched. Needles and plungers must be replaced as a set
Specifications
Specifications
| Material | Stainless Steel, Stainless Steel (Tip) |
| Needle Gauge | 23 G |
| Diameter (Metric) Outer | 0.63 mm |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Includes | Matched plunger and needle |
| Product Type | Replacement Needle and Plunger: NP0.5BN-5/0.63C |
| Volume (Metric) | 0.5 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | 6.5 mm OD Syringe (0.5μL) |