Learn More
Trajan™ Replacement Ferrules
Click to view available options
:
For Capillary Tubing 0.1 to 0.25mm I.D.
Description
- Composite of graphite and Vespel
- Mechanically robust, hard material, long lifetime
- Forms strong grip with capillary column
- Suitable for Thermo Scientific instruments
Specifications
Specifications
| Type | Replacement Ferrule |
| For Use With (Equipment) | Capillary Tubing 0.1 to 0.25 mm I.D., Use for Mini Unions |