Learn More
Trajan™ PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing. Trajan Scientific and Medical™ PEEKsil™ Tubing can be used for sealing with conventional metal or polymer ferrule systems.
Brand:  Trajan™ 0624350
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15298773
  • 755.00 NOK / Pack of 2
Estimated Shipment: 03-07-2024
Description
  • Robust - PEEK polymer exterior and fused silica inner tubing
  • Smooth wall surface giving excellent flow characteristics
  • Compatible with most organic solvents
  • Inertness - extremely low absorption characteristics
  • Precise ID
Specifications
Specifications
Tubing
0.0625 in.
500 mm
1.5 mm
0.175 mm