Learn More
Trajan™ Carlo Erba On-Column Syringe
Product Code. 10626231
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10626231 5 μL Each
10015510 10 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10626231 Supplier Trajan™ Supplier No. 000380
Estimated Shipment: 17-11-2026
On-column syringes require the needle OD to be smaller than the ID of the capillary column with which it is used. For use with Carlo Erba products
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Designed for on-column injections
Specifications
Diameter (Metric) Outer 0.23 mm
Product Type On Column Syringe
Volume (Metric) 5 μL
Length (Metric) Needle 75 mm