Learn More
SGE™ NanoVolume Syringes
Product Code. 10725046
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
Needle Gauge:
23 G
26 G
Model:
0.5BNR-5
0.5BNR-5-RAX
0.5BNR-5/0.47
0.5BNR-5BV
0.5BR-7
0.5BR-7-RAX
0.5BR-7BV
0.5BR-OC-7/0.47
1BR-11.5
1BR-5
1BR-5-RAX
1BR-5BV
1BR-7
1BR-7/0.47
1BR-7BV
5BR-11.5
5BR-5
5BR-5BV
5BR-7
5BR-7-RAX
5BR-7BV
Needle Point Style:
Cone
Removable Bevel
Removable Cone
Unit Size:
Each
Product Code. Volume (Metric) Needle Gauge Model Needle Point Style unitSize
10725046 1 μL 23 G 1BR-5BV Removable Bevel Each
10711432 0.5 μL 23 G 0.5BNR-5 Removable Cone Each
10370901 0.5 μL 26 G 0.5BNR-5/0.47 Removable Cone Each
10482001 0.5 μL 23 G 0.5BR-7 Removable Cone Each
11982585 0.5 μL 23 G 0.5BR-7BV Removable Bevel Each
10306392 0.5 μL 23 G 0.5BR-7-RAX Removable Cone Each
10095500 0.5 μL 23 G 0.5BNR-5-RAX Removable Cone Each
10636231 0.5 μL 26 G 0.5BR-OC-7/0.47 Removable Bevel Each
10616421 1 μL 23 G 1BR-7 Removable Cone Each
11992585 1 μL 23 G 1BR-7BV Removable Bevel Each
10167260 1 μL 23 G 1BR-11.5 Removable Cone Each
10568882 1 μL 26 G 1BR-7/0.47 Removable Cone Each
10601813 1 μL 23 G 1BR-5-RAX Removable Cone Each
10318983 5 μL 23 G 5BR-5BV Removable Bevel Each
10588882 5 μL 23 G 5BR-7 Removable Cone Each
11902595 5 μL 23 G 5BR-7BV Removable Bevel Each
12907536 5 μL 23 G 5BR-11.5 Cone Each
10578302 5 μL 23 G 5BR-7-RAX Cone Each
11962585 0.5 μL 23 G 0.5BNR-5BV Removable Bevel Each
10763714 1 μL 23 G 1BR-5 Removable Cone Each
10288860 5 μL 23 G 5BR-5 Removable Cone Each
21 options
Product Code. 10725046
Brand: SGE™ 000501
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
  • Ability to inject down to 50nL with high precision and accuracy
  • Ideal for dispensing very small liquid volumes
  • Plunger extends into the needle tip
  • Sample is contained only in the needle without any glass contact
  • Virtually zero dead volume and carry over between injections
  • Recommended for use when sample volume is less than 1000nL (1μL)
Specifications
Accuracy ±2%
Material Borosilicate Glass (Barrel)
Needle Gauge 23 G
Diameter (English) Outer Needle 0.0248 in.
Diameter (English) Outer 0.315 in.
Diameter (Metric) Outer 8 mm
Diameter (Metric) Outer Needle 0.63 mm
Length (English) 2 in.
Length (English) Needle 1.96 in.
Length (Metric) 50 mm
Product Type NanoVolume Syringe
Model 1BR-5BV
Quantity Ea.
Volume (Metric) 1 μL
Needle Point Style Removable Bevel
Connection Type Removable