Learn More
SGE™ MEPS™ Syringes
Description
- The eVol™ custom programming function enables MEPS to be semi-automated and proofed before transition to fully automated platforms
Specifications
Specifications
| Diameter (English) Inner | 0.26 in. |
| Diameter (Metric) Inner | 6.7 mm |
| For Use With (Application) | Gas/Liquid chromatography |
| Product Type | MEPS™ Syringe |
| Quantity | Ea. |
| Volume (Metric) | 100 μL |
| For Use With (Equipment) | CTC Analytics, HTA 300APlus, Thermo Scientific and Varian 8400 System |
| Connection Type | Removable |