Learn More
Trajan™ GC Capillary Column Connectors: Replacement μ-Unions
Product Code. 15248943
Change view
Click to view available options
Connections:
0.1 - 0.32 mm and 0.53 mm
0.1 to 0.32 mm
0.53 mm and 0.53 mm
Unit Size:
Pack of 5
Product Code. Connections unitSize
15248943 0.1 - 0.32 mm and 0.53 mm Pack of 5
15258943 0.1 to 0.32 mm Pack of 5
15278943 0.53 mm and 0.53 mm Pack of 5
3 options
Product Code. 15248943
Brand: Trajan™ 073573
Only null left
Add to basket
Add to basket
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type Replacement μ-Union
Connections 0.1 - 0.32 mm and 0.53 mm
For Use With (Equipment) GC Capillary Column Connectors