Learn More
Trajan™ GC Capillary Column Connectors: Replacement Ferrules
Product Code. 15298933
Click to view available options
For Use With (Equipment):
0.1 to 0.25 mm I.D.
0.1 to 0.25 mm I.D., 0.32 mm I.D.
0.1 to 0.25 mm I.D., 0.53 mm I.D.
0.32 mm I.D.
0.32 mm I.D., 0.53 mm I.D.
0.53 mm I.D.
Unit Size:
Pack of 10
Product Code. 15298933
Brand: Trajan™ undefined
Only null left
Add to basket
Add to basket
Replacement SilTite ferrules for SilTiteμ-Union
  • SilTite μ-Union gives low thermal mass - 9mm in length and <0.5g mass
  • Zero dead volume for optimized peak shape
  • Chemically deactivated stainless steel ferrules give system inertness
  • Robust and stable for both high temperature and pressure cycling
Specifications
Type Replacement μ-Union Ferrule
Material Stainless Steel
For Use With (Equipment) 0.1 to 0.25 mm I.D., 0.32 mm I.D.