Learn More
SGE™ Gas Tight Syringe with Removable Needle
Product Code. 10163352
Change view
Click to view available options
Length (English):
1.65 in.
1.96 in.
Length (English) Needle:
1.65 in.
1.96 in.
Model:
10R-S-GT-0.63C
250R-V-GT
Needle Point Style:
Fixed Bevel
Removable Cone
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Length (English) Needle Model Needle Point Style unitSize
10163352 1.96 in. 1.96 in. 250R-V-GT Fixed Bevel Each
10494422 1.65 in. 1.65 in. 10R-S-GT-0.63C Removable Cone Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10163352 Supplier SGE™ Supplier No. 006279
Only null left
Add to basket
Add to basket
Ideal for sample storage, transportation and pressurizing samples prior to injection
  • Removable needle
  • Gas tight plunger
  • Stainless steel
  • Gauge: 23
Specifications
Needle Gauge 23 G
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Ideal for sample storage and transportation or to pressurize sample prior to injection
Length (English) 1.96 in.
Length (English) Needle 1.96 in.
Length (Metric) 50 mm
Product Type Analytical Syringe
Model 250R-V-GT
Quantity Ea.
Volume (Metric) 250 μL
Length (Metric) Needle 50 mm
Needle Point Style Fixed Bevel
Connection Type Removable