Learn More
SGE™ GC Capillary Columns: Phenyl (equiv.) Polycarborane Siloxane
Click to view available options
Length (Metric):
12 m
15 m
25 m
30 m
Diameter (Metric) Inner:
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.15 μm
0.5 μm
Material:
Polyimide Clad
Unit Size:
Each
Description
- Low bleed; high inertness for most demanding analyses
- High tolerance level to adverse conditions and samples
- Manufactured to exact specifications, ensuring column-to-column reproducibilty
- forte HT5 (5% Phenyl Polycarborane-siloxane)—High 480°C temperature phase suited for simulated distillation and other petroleum applications
Specifications
Specifications
| Max. Temperature | 400°C |
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | 5% Phenyl (equiv.) Polycarborane Siloxane |
| For Use With (Application) | Simulated Distillation, Pesticide Analysis, Herbicides Analysis |
| Particle Size | 0.1 μm |
| Temperature Range | 10°C to 380°C |
| Film Thickness | 0.1 μm |
| Length (Metric) | 15 m |
| Material | Polyimide Clad |
| Packing Material | HT5 |
| Show More |