Learn More
SGE™ BP20 (Wax) GC Capillary Columns
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
2 μm
Unit Size:
Each
Description
Features
- Industry standard Wax column
- Bonded and cross-linked
- Can be solvent rinsed
Applications
- Used for separation of Alcohol, Free acids, Fatty acids methyl esters, Aromatics, Solvents and Essential oils
Specifications
Specifications
| Max. Temperature | 280°C |
| Functionality/Interaction Type | Hydrogen Bonding or Dipole interactions |
| Diameter (Metric) Inner | 0.32 mm |
| Stationary Phase | SGE™ BP20 (Wax) |
| USP Type | G14, G15, G16, G20, G39 |
| For Use With (Application) | Free Acids Analysis, Free Acids, Fatty Acid Methyl Esters, Aromatics Analysis, Solvents and Essential Oils |
| Temperature Range | 20°C to 260/280°C |
| Film Thickness | 0.25 μm |
| Length (Metric) | 60 m |
| Polarity | Polar |
| Show More |