Learn More
Trajan™ BP1 Pona GC Capillary Columns
Click to view available options
Length (Metric):
50 m
Unit Size:
Each
Description
- 100% Dimethyl Polysiloxane
- Designed for the analysis of petroleum products
- Non-polar phase for PONA analysis
Specifications
Specifications
| Max. Temperature | 320/340°C |
| Diameter (Metric) Inner | 0.15 mm |
| Film Thickness | 0.5 μm |
| Length (Metric) | 50 m |