Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Product Code. 12927586
Change view
Click to view available options
Volume (Metric):
20 to 500 μL
5 μL
50 μL
500 μL
Needle Point Style:
Bevel
Without Needle
Quantity:
3/Pk.
Ea.
Unit Size:
Each
Pack of 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Point Style Quantity Unit Size
12927586 5 μL Bevel Ea. Each
12997586 500 μL Bevel Ea. Each
12947586 50 μL Bevel Ea. Each
12907596 20 to 500 μL Without Needle Ea. Each
12927596 5 μL Bevel 3/Pk. Pack of 3
12937596 50 μL Bevel 3/Pk. Pack of 3
12957596 500 μL Bevel 3/Pk. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12927586 Supplier SGE™ Supplier No. 2910020
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 13-10-2026
Improves laboratory workflow
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
  • Digitally controlled positive displacement dispensing system
  • Programmable—enables user-independent liquid handling procedures
  • Easily calibrated to ensure accurate dispensing
  • Stores calibration factors for up to ten XCHANGE™ syringes
  • Touch Wheel user interface
  • Full color screen
Applications:
  • Preparation of calibration standards
  • Addition of internal standards
  • Precise dispensing of non-aqueous liquids
  • Sample dilution
Specifications
Needle Gauge 25 G
Diameter (English) Inner Needle 0.0047 in.
Diameter (Metric) Inner Needle 0.12 mm
Diameter (English) Outer Needle 0.019 in.
Diameter (Metric) Outer Needle 0.5 mm
For Use With (Application) Preparation of calibration standards, addition of internal standards
Length (English) Needle 1.96 in.
Product Type eVol Analytical Syringe
Quantity Ea.
Volume (Metric) 5 μL
Needle Point Style Bevel
For Use With (Equipment) eVol Automated Syringe