Learn More
Trajan™ Diamond MS Manual Syringes
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Model:
100R-GT-MS1
10R-GT-MS1
250R-GT-MS1
25R-GT-MS1
500R-GT-MS1
50R-GT-MS1
Unit Size:
Each
Description
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
| Includes | Removable ultra smooth needle |
| Product Type | Diamond MS Manual Syringe |
| Model | 100R-GT-MS1 |
| Quantity | Ea. |
| Volume (Metric) | 100 μL |
| Connection Type | Removable |