Learn More
Trajan™ Diamond LC Needles for 100 to 500μL Manual and Autosampler Syringes
Product Code. 13127916
Change view
Click to view available options
Diameter (Metric) Inner:
0.17 mm
0.37 mm
Length (English) Needle:
2 in.
2.24 in.
Includes:
Ultra Smooth Coating
Ultra Smooth Hydrophilic Coating
Ultra Smooth Hydrophobic Coating
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (English) Needle Includes unitSize
13127916 0.37 mm 2 in. Ultra Smooth Coating Each
13127906 0.37 mm 2 in. Ultra Smooth Hydrophilic Coating Each
13117906 0.17 mm 2 in. Ultra Smooth Hydrophilic Coating Each
13167906 0.17 mm 2 in. Ultra Smooth Hydrophobic Coating Each
13147906 0.17 mm 2.24 in. Ultra Smooth Hydrophilic Coating Each
13177906 0.37 mm 2 in. Ultra Smooth Hydrophobic Coating Each
13197906 0.17 mm 2.24 in. Ultra Smooth Hydrophobic Coating Each
13117916 0.17 mm 2 in. Ultra Smooth Coating Each
13147916 0.17 mm 2.24 in. Ultra Smooth Coating Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 13127916 Supplier Trajan™ Supplier No. 0355233
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes are MS ready and designed to improve the sensitivity of your MS results.
Improve the sensitivity of your MS analysis:
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Material Stainless Steel
Needle Gauge 22 G
Diameter (English) Inner 0.014 in.
Diameter (English) Outer 0.02 in.
Diameter (Metric) Inner 0.37 mm
Diameter (Metric) Outer 0.72 mm
Includes Ultra Smooth Coating
Length (English) Needle 2 in.
Product Type Diamond Needle
Needle Point Style LC
For Use With (Equipment) Manual and Autosampler Syringes (100 to 500μL)