Learn More
Trajan™ Diamond CTC PAL 100μL GC Autosampler Syringes
Product Code. 13167936
Click to view available options
Needle Point Style:
Ultra Smooth Hydrophilic, Cone
Ultra Smooth Hydrophobic, Cone
Ultra Smooth, Cone
Unit Size:
Each
Product Code. 13167936
Brand: Trajan™ 0355521
Improves sensitivity of MS Analysis with reduced non specific sample interaction and increased analyte recovery.
  • Ensures carryover is reduced to levels lower even than the best fixed needle syringes available. In the event of needle damage, Diamond MS needles provide a cost effective alternative to purchasing an entire fixed needle syringe.
  • Ultra Smooth needles reduce non specific interaction between your sample and the needle. 
  • Further reduce sample interactions with a hydrophobic or hydrophillic needle coating
Specifications
Needle Gauge 23 G
Diameter (English) Inner Needle 0.009 in.
Diameter (Metric) Inner Needle 0.24 mm
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Length (English) Needle 1.9 in.
Product Type Autosampler Syringe
Quantity 1 Pk.
Volume (Metric) 100 μL
Needle Point Style Ultra Smooth Hydrophobic, Cone
For Use With (Equipment) CTC PAL GC