Learn More
Trajan™ CTC LC Autosampler Syringes
Product Code. 12907526
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Each
Product Code. 12907526
Brand: Trajan™ 002710
Borosilicate glass barrel and stainless steel needles provide strength and inertness. Backing provides quick and accurate volume assessment.
  • High temperature syringes for headspace application with temperatures up to 150°C
  • PTFE plunger seal option for smooth, leak free operation - suitable for both liquid and gas samples, with replaceable plungers
  • Specifications of the syringes are exactly matched to CTC holders to ensure proper fit and reliable functioning
Specifications
Needle Gauge 22 G
Diameter (English) Outer Needle 0.028 in.
Diameter (Metric) Outer Needle 0.72 mm
For Use With (Application) High performance liquid chromatology
Length (English) Needle 2 in.
Product Type Autosampler Syringe
Model 10F-CTC-LC
Quantity Ea.
Volume (Metric) 10 μL
Length (Metric) Needle 51 mm
Needle Point Style LC
Connection Type Fixed