Learn More
SGE™ BPX70 Capillary Columns
Description
Specifically engineered for the analysis of fatty acid methyl esters (FAMEs)
- The only high-temperature, high cyanopropyl content stationary phase used routinely for GC/MS applications
- (70%)-Cyanopropyl (equiv.) polysilphenylene-siloxane
Suggested Primary Applications for BPX70 Columns:
Fatty Acid Methyl Esters (FAMEs)
Pharmaceuticals
Sugars
Specifications
Specifications
| Diameter (Metric) Inner | 0.32 mm |
| Stationary Phase | SGE™ BPX70 |
| For Use With (Application) | Carbohydrate Analysis, Carbohydrates, Pharmaceuticals Analysis |
| Particle Size | 0.25 μm |
| Temperature Range | 50°C to 250/260°C |
| Film Thickness | 0.25 μm |
| Length (Metric) | 25 m |
| Polarity | Polar |