Learn More
Trajan™ GC/MS Interface Connections for Varian GC Instruments
Product Code. 12947101
Change view
Click to view available options
For Use With:
0.10 to 0.25 mm Column
0.32 mm ID Column
0.45 to 0.53 mm ID Columns
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
12947101 0.45 to 0.53 mm ID Columns Each
12937101 0.32 mm ID Column Each
12634945 0.10 to 0.25 mm Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12947101 Supplier Trajan™ Supplier No. 073302
Only null left
Add to basket
Add to basket
Delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type Ferrule
Diameter (Metric) Inner 0.45 to 0.53 mm column
Packaging Quantity 10 ferrules, 2 nuts
For Use With 0.45 to 0.53 mm ID Columns
Material SilTite