Learn More
SGE™ BP1 Capillary Columns
Product Code. 15248713
Click to view available options
Length (Metric):
10 m
15 m
30 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
1 μm
Unit Size:
Each
Product Code. 15248713
Brand: SGE™ 054815
Only null left
Add to basket
Add to basket
Industry-standard nonpolar phase suitable for all routine analyses
  • 100% dimethylpolysiloxane
  • Extremely low-bleed phase with a high temperature limit of 320/340°C
Specifications
Diameter (Metric) Inner 0.25 mm
Stationary Phase SGE™ BP1
For Use With (Application) Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
Particle Size 0.25 μm
Temperature Range -60°C to +340/360°C
Film Thickness 0.25 μm
Length (Metric) 60 m
Polarity Non polar