Learn More
Trajan™ Autosampler Syringes
Product Code. 10380611
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
10 μL
Needle Gauge:
23 G
25 G
26 G
Connection Type:
Fixed
Plunger-In, Repeating Adapter
Removable
For Use With (Equipment):
Carlo Erba/Fisons/Leap/CTC AS200 and AS800, GC autosampler syringe
Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus
PerkinElmer Autosystem
Shimadzu AOC 14, 17, 20 and 20i GCs
Varian 8035, 8100 and 8200 GC Autosamplers
Unit Size:
Each
Product Code. Volume (Metric) Needle Gauge Connection Type For Use With (Equipment) unitSize
10380611 10 μL 23 G Removable Shimadzu AOC 14, 17, 20 and 20i GCs Each
10214942 1 μL 23 G Plunger-In, Repeating Adapter - Each
10452561 0.5 μL 23 G Removable Shimadzu AOC 14, 17, 20 and 20i GCs Each
10611813 5 μL 26 G Fixed PerkinElmer Autosystem Each
10143402 5 μL 23 G Fixed PerkinElmer Autosystem Each
10033662 5 μL 26 G Fixed Carlo Erba/Fisons/Leap/CTC AS200 and AS800, GC autosampler syringe Each
10245799 5 μL 23 G Fixed Shimadzu AOC 14, 17, 20 and 20i GCs Each
10577922 10 μL 26 G Removable Shimadzu AOC 14, 17, 20 and 20i GCs Each
10316772 10 μL 25 G Removable Varian 8035, 8100 and 8200 GC Autosamplers Each
10431031 10 μL 26 G Fixed Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus Each
15208473 10 μL 26 G Removable Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus Each
11 options
Product Code. 10380611
Brand: Trajan™ 002898
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
Needle Gauge 23 G
Diameter (English) Outer Needle 0.02 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Length (English) 1.65 in.
Length (English) Needle 1.6 in.
Length (Metric) 42 mm
Product Type Autosampler Syringe
Model 10R-S-0.63C
Quantity Ea.
Volume (Metric) 10 μL
Needle Point Style Cone
For Use With (Equipment) Shimadzu AOC 14, 17, 20 and 20i GCs
Connection Type Removable