Learn More
Abnova™ SGCA (Human) Recombinant Protein
Product Code. p-7164647
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16122708
Brand: Abnova™ H00006442P01.10ug
Human SGCA full-length ORF ( NP_000014.1, 1 a.a. - 387 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MAETLFWTPLLVVLLAGLGDTEAQQTTLHPLVGRVFVHTLDHETFLSLPEHVAVPPAVHITYHAHLQGHPDLPRWLRYTQRSPHHPGFLYGSATPEDRGLQVIEVTAYNRDSFDTTRQRLVLEIGDPEGPLLPYQAEFLVRSHDAEEVLPSTPASRFLSALGGLWEPGELQLLNVTSALDRGGRVPLPIEGRKEGVYIKVGSASPFSTCLKMVASPDSHARCAQGQPPLLSCYDTLAPHFRVDWCNVTLVDKSVPEPADEVPTPGDGILEHDPFFCPPTEAPDRDFLVDALVTLLVPLLVALLLTLLLAYVMCCRREGRLKRDLATSDIQMVHHCTIHGNTEELRQMAASREVPRPLSTLPMFNVHTGERLPPRVDSAQVPLILDQH
Specifications
Accession Number NP_000014.1
Gene ID (Entrez) 6442
Name sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein)
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias 50-DAG, A2, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin
Gene Symbol SGCA
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer