missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS17A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13441-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
DXYS155E is a gene found in the pseudoautosomal region of the distal short arms of the X and Y chromosomes, and appears to be ubiquitously expressed.Specifications
SFRS17A | |
Polyclonal | |
Unconjugated | |
AKAP17A | |
721P, B lymphocyte antigen, CCDC133, CXYorf3, DXYS155E, Protein XE7, SFRS17A, XE7, XE7Y | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8227 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q02040 | |
AKAP17A | |
SFRS17A Fusion Protein Ag4252 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |