Learn More
SF3B2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10919-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
SF3b is a U2 snRNP-associated protein complex essential for spliceosome assembly. SF3b contains the spliceosomal proteins SAP 49, SAP 130, SAP 145, and SAP 155. SAP 130, SAP 145, and SAP 155 are present in a protein complex in HeLa nuclear extracts and associate with one another. While SAP 155 and SAP 130 interact with each other (directly or indirectly) within this complex, SAP 49 and SAP 145 are known to interact directly with each other. Unexpectedly, the SAP 49-SAP 145 protein-protein interaction requires the amino-terminus of SAP 49, which contains two RNA-recognition motifs. The observation that SAP 49 and SAP 145 interact directly with both U2 snRNP and the pre-mRNA suggests that this protein complex plays a role in tethering U2 snRNP to the branch site.Specifications
SF3B2 | |
Polyclonal | |
Unconjugated | |
SF3B2 | |
SAP 145, SAP145, SF3B145, SF3b150, SF3B2, Splicing factor 3B subunit 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10992 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13435 | |
SF3B2 | |
SF3B2 Fusion Protein Ag1009 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |