Learn More
SF3B1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27684-1-AP-20UL
 View more versions of this product
Product Code. 16845775
  • 1710.33 NOK / 20µL
Description
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.
Specifications
Specifications
SF3B1
Polyclonal
Unconjugated
SF3B1
PRP10, PRPF10, SAP 155, SAP155, SF3B1, SF3b155, Splicing factor 3B subunit 1
Rabbit
Antigen Affinity Chromatography
RUO
23451
-20°C
Liquid
Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75533
SF3B1
SF3B1 Fusion Protein Ag26689