Learn More
SF1670, MedChemExpress
Product Code. 16473496
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16473496
Brand: Medchem Express HY15842
MedChemExpress SF1670 is a potent and specific phosphatase and tensin homolog deleted on chromosome 10 (PTEN) inhibitor.
SF1670 is a potent and specific phosphatase and tensin homolog deleted on chromosome 10 (PTEN) inhibitor.
Specifications
Chemical Name or Material SF1670
Physical Form Solid
CAS 345630-40-2
Color Red
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C19H17NO3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 50 mg/mL (162.69 mM)
SMILES CC(C)(C)C(NC(C=C1C2=O)=CC=C1C3=C(C2=O)C=CC=C3)=O
Molecular Weight (g/mol) 307.34
Formula Weight 307.34
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.