Learn More
Seward™ Stomacher™ Simplette™ Pipets
Product Code. 11830933
Change view
Click to view available options
Packaging Quantity:
2000/Pk.
Unit Size:
Pack of 2000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
11830933 2000/Pk. Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11830933 Supplier Seward™ Supplier No. BA6051
  • ONLINE EXCLUSIVE PRICE
    542.00 NOK
    503.10 NOK / Pack of 2000
    Save 38.90 NOK
    7% Off
Estimated Shipment: 02-12-2026
Seward Simplettes™ provide an accurate and economical alternative to pipette tips, designed specifically to pipette samples from the bottom of the Stomacher™ bag without risk of cross contamination.
Helps facilitate sample transfer; Are of the right length to reach down to the bottom of the bag to recover the sample for inoculating Petri dishes and broths; Come in 2 sizes standard and long; Capable of pipetting volumes between 0.1ml and 2ml; irradiated sterile
Specifications
Product Type Transfer Pipet
Diameter (Metric) 4.1 mm
Length (Metric) 177 mm
Description Simplettes; Transfer Pipet
For Use With (Application) Ideal to pipette samples from sample bag
For Use With (Equipment) All Stomacher sample bags
Packaging Quantity 2000/Pk.