Learn More
Seward™ Stomacher™ Bag Racks
Description
Help to organize sample bags
- High-grade plastic polymer construction
- Provide perfect storage for Stomacher bag sample handling for a cleaner, more productive work environment
- Perfect for loading closure bags pre-enrichment into the incubator
- Easily dismantled and cleaned with most lab disinfectants (not autoclavable)
Specifications
Specifications
| Description | Sample bag rack (10 places) |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Holds | Bags |
| Material | Polymer |
| Positions | 10 |
| Sterility | Non-sterile |
| For Use With (Application) | To prepare, store and transport samples in microbiology sample preparation |
| For Use With (Equipment) | Stomacher 80Biomaster |
| No. per Pack | 1 |
| Show More |