Learn More
Cole-Parmer™ Settleometer
Product Code. 15385323 Shop All Cole-Parmer Products
Change view
Click to view available options
Volume (Metric):
1.4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15385323 1.4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15385323 Supplier Cole-Parmer™ Supplier No. 0547600
  • ONLINE EXCLUSIVE PRICE
    1475.00 NOK
    1395.53 NOK / Pack of 1
    Save 79.47 NOK
    5% Off
Estimated Shipment: 29-12-2026
Assists wastewater treatment plant operators in performing sludge quality control tests.
  • Measure the rate and amount of solid/liquid separation of sludge samples
  • Easy-to-read white calibration markings indicate percent solid and settled sludge volume
Specifications
Description Settlometer Kit, acrylic, 1.4 L, dual scale % solids and SSV
Volume (Metric) 1.4 L
Includes Mixing paddle, Snap-on lid, and Instructions
Material Acrylic