missing translation for 'onlineSavingsMsg'
Learn More

SETDB1 Mouse anti-Human, Clone: 1H6E5, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66293-1-IG-150UL

 View more versions of this product

Product Code. 16848035

  • 4836.10 NOK / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N (6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.
Specifications

Specifications

SETDB1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15047
SETDB1
SETDB1 Fusion Protein Ag21644
150 μL
Primary
Human
Antibody
IgG1
Western Blot
1H6E5
Unconjugated
SETDB1
ESET, H3 K9 HMTase 4, H3 K9 HMTase4, KG1T, KIAA0067, KMT1E, Lysine N methyltransferase 1E, SET domain bifurcated 1, SET domain, bifurcated 1, SETDB1
Mouse
Protein G
RUO
9869
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.