missing translation for 'onlineSavingsMsg'
Learn More

SETBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16841-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16846604

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein which contains a several motifs including a ski homology region and a SET-binding region in addition to three nuclear localization signals. The encoded protein has been shown to bind the SET nuclear oncogene which is involved in DNA replication. Mutations in this gene are associated with Schinzel-Giedion midface retraction syndrome. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SETBP1
Polyclonal
Unconjugated
SETBP1
KIAA0437, SEB, SET binding protein, SET binding protein 1, SETBP1
Rabbit
Antigen Affinity Chromatography
RUO
240427, 26040, 291423
-20°C
Liquid
Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y6X0, Q9Z180
SETBP1
SETBP1 Fusion Protein Ag10532
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers