Learn More
Interscience™ Set of Tygon™ Tubings for Pump Head
Description
- Transparent
- Flexible Tygon™ tubing
Specifications
Specifications
| Product Type | Pump Tubing |
| Autoclavable | Autoclavable |
| For Use With (Application) | Adding Tubing to Dilutor Pumps |
| For Use With (Equipment) | DiluFlow, DiluFlow Pro, DiluFlow Elite (5 kg), FlexiPump, FlexiPump Pro |
| Quantity | 6 Tubings/Set |
| Diameter (Metric) Inner | 4.8 mm |