Learn More
Interscience™ Set of Tygon™ Tubings for Pump Head
Product Code. 17860087
Change view
Click to view available options
Diameter (Metric) Inner:
4.8 mm
6.4 mm
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
17860087 6.4 mm Pack of 6
12818291 4.8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17860087 Supplier Interscience™ Supplier No. 561304
  • ONLINE EXCLUSIVE PRICE
    767.00 NOK
    743.70 NOK / Pack of 6
    Save 23.30 NOK
    3% Off
 Restricted Item
Estimated Shipment: 20-01-2027
20 cm long Tygon™ tubings for use with DiluFlow and FlexiPump pump heads.
  • Transparent
  • Flexible Tygon™ tubing
Specifications
Product Type Pump Tubing
Autoclavable Autoclavable
For Use With (Application) Adding Tubing to Dilutor Pumps
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro
Quantity 6 Tubings/Set
Diameter (Metric) Inner 6.4 mm