Learn More
Set Diesel Oil and Mineral Oil, Dr. Ehrenstorfer
Product Code. 30312615
Change view
Click to view available options
Quantity:
2 x (3 x 1 mL)
Unit Size:
6mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312615 2 x (3 x 1 mL) 6mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30312615 Supplier Dr. Ehrenstorfer Supplier No. DRECA03009020
 Restricted Item
Estimated Shipment: 30-11-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material No. 2 Diesel Oil
CAS 68334-30-5
Analyte or Component Names No. 2 Diesel Oil
Shipping Condition Room Temperature
Certifications/Compliance ISO/IEC 17025
Synonym Fuels, diesel, Diesel fuel oil, Diesel oil, Diesel oil (petroleum), Diesel test fuel, HRD 76, NATO-F 76, Diesel fuel
Solution Type Neat
Industry Type Environmental Testing
Quantity 2 x (3 x 1 mL)