Learn More
Interscience™ Set of 5 Tube Weights
Product Code. 15774057
Change view
Click to view available options
Quantity:
5 Weights/Set
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15774057 5 Weights/Set Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15774057 Supplier Interscience™ Supplier No. 513039
  • ONLINE EXCLUSIVE PRICE
    2745.00 NOK
    2611.26 NOK / Pack of 5
    Save 133.74 NOK
    4% Off
Estimated Shipment: 14-12-2026
Set of 5 tube weights for use with dilutors.
  • For tubing with 6 mm diameter
Specifications
Product Type Tube Weight
Autoclavable Autoclavable
For Use With (Application) Applying Weights to Tubing
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro
Quantity 5 Weights/Set